Learn More
Aviva Systems Biology Corp HOXC9 Antibody - middle region (ARP35813_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35813_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXC9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human HOXC9. Alternate names: HOX3, HOX3BApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-55870