Learn More
Aviva Systems Biology Corp HOXD1 Antibody - C-terminal region (ARP32644_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32644_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HOXD1. Alternate names: HOX4, HOX4G, Hox-4.7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54687