Learn More
Aviva Systems Biology Corp HOXD1 Antibody - middle region (ARP57870_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57870_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXD1. Alternate names: HOX4, HOX4G, Hox-4.7ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62166