Learn More
Aviva Systems Biology Corp HOXD10 Antibody - middle region (P100936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXD10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXD10. Alternate names: HOX4, HOX4D, HOX4E, Hox-4.4ApplicationWB. Species reactivity: Cow, Dog, Horse, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62789