Learn More
Aviva Systems Biology Corp HOXD12 Antibody - N-terminal region (ARP32647_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32647_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXD12. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HOXD12. Alternate names: HOX4HApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54688