Learn More
Aviva Systems Biology Corp HOXD3 Antibody - C-terminal region (ARP37883_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37883_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse HOXD3. Alternate names: Hox-4.1, Hox-5.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56456