Learn More
Aviva Systems Biology Corp HOXD3 Antibody - middle region (ARP31964_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31964_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human HOXD3. Alternate names: HOX1D, HOX4, HOX4A, Hox-4.1, MGC10470ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52847