Learn More
Aviva Systems Biology Corp HOXD4 Antibody - C-terminal region (ARP31391_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31391_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HOXD4. Alternate names: HOX4, HOX4B, HHO.C13, HOX-5.1, Hox-4.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52824