Learn More
Neta Scientific HP-1 25M 0.32MM 0.17U
SDP
Catalog No. 50928812
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-812 Supplier Neta Scientific Supplier No. 19091Z012
This item is manufactured by Agilent