Learn More
Optimize Technologies OPTI-MAX EXP Single Ball Upgrade for UHPLC Pressures, 1/8" Single Ceramic Ball & Seat, SS Replacement Cartridge with PEEK endcaps, Fits all OPTI-MAX Housings (except xx-6x-xxxxx) 2pk
SDP
Catalog No. 502006208
Encompass_Preferred
Opti-Max Check Valves are manufactured with ball and seat sets that are individually matched to ensure a lock-and-key fit. This guarantees 3 bubbles per minute or less in the standard nitrogen bubble test. Optimize has also minimized the ball travel distance within the check valve which results in quicker seating and more accurate pump metering. A single OPTI-MAX cartridge is interchangeable across multiple pump brands, and will function in either the inlet or outlet position. The same replacement cartridge will fit into OPTI-MAX housings for pumps made by Waters, Shimadzu, Agilent/HP, and most other OEM solvent delivery systems. This allows you to minimize the number of spare check valve cartridges you keep on hand, and dramatically reduce the cost of inventory required to cover your maintenance needs. This particular check valve is recommended for acetonitrile applications. Please contact your Fisher or Optimize representative to view a full list of OPTI-MAX cartridges.
Catalog No. 50-200-6208 Supplier Optimize Technologies Supplier No. 109602004
Opti-Max Check Valves are manufactured with ball and seat sets that are individually matched to ensure a lock-and-key fit. This guarantees 3 bubbles per minute or less in the standard nitrogen bubble test. Optimize has also minimized the ball travel distance within the check valve which results in quicker seating and more accurate pump metering. A single OPTI-MAX cartridge is interchangeable across multiple pump brands, and will function in either the inlet or outlet position. The same replacement cartridge will fit into OPTI-MAX housings for pumps made by Waters, Shimadzu, Agilent/HP, and most other OEM solvent delivery systems. This allows you to minimize the number of spare check valve cartridges you keep on hand, and dramatically reduce the cost of inventory required to cover your maintenance needs. This particular check valve is recommended for acetonitrile applications. Please contact your Fisher or Optimize representative to view a full list of OPTI-MAX cartridges.