Learn More
Grainger Zoeller Effluent Pump, 1 HP, Cast Iron, 115V AC, 1 ea
Catalog No. 502781934
Encompass_Preferred
The Model 140 Effluent Pump is a robust 1 HP corded electric pump designed for efficient effluent water handling. Featuring a durable, powder-coated epoxy cast iron construction, this pump is built for longevity and corrosion resistance in demanding environments. It includes a piggyback tether float switch for automatic operation and a vortex impeller design to manage spherical solids.
  • 1 HP motor with 3,450 RPM speed
  • 115 V AC, 60 Hz operation with 12 A current draw
  • Maximum flow rate of 86 gpm and shut-off head of 50 ft
  • Continuous duty cycle with automatic thermal overload protection
  • Discharge size of 1-1/2 in. NPT
  • Capable of handling 3/4 in. spherical solids
  • Includes 20 ft. (6 m) SOW power cord
  • Certified to CSA standards
Catalog No. 50-278-1934 Supplier Grainger Supplier No. 19T426
The Model 140 Effluent Pump is a robust 1 HP corded electric pump designed for efficient effluent water handling. Featuring a durable, powder-coated epoxy cast iron construction, this pump is built for longevity and corrosion resistance in demanding environments. It includes a piggyback tether float switch for automatic operation and a vortex impeller design to manage spherical solids.
  • 1 HP motor with 3,450 RPM speed
  • 115 V AC, 60 Hz operation with 12 A current draw
  • Maximum flow rate of 86 gpm and shut-off head of 50 ft
  • Continuous duty cycle with automatic thermal overload protection
  • Discharge size of 1-1/2 in. NPT
  • Capable of handling 3/4 in. spherical solids
  • Includes 20 ft. (6 m) SOW power cord
  • Certified to CSA standards