Learn More
Neta Scientific HP-5 17M 0.20MM 0.33UM
SDP
Catalog No. 50928637
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-637 Supplier Neta Scientific Supplier No. 19091J108
This item is manufactured by Agilent