Learn More
Neta Scientific HP-5 30M 0.32MM 0.5U
SDP
Catalog No. 50928863
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-928-863 Supplier Neta Scientific Supplier No. 19091J113
This item is manufactured by Agilent