Learn More
Neta Scientific HP-CHIRAL-20B 30M 0.32MM 0.2
SDP
Catalog No. 50927212
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-927-212 Supplier Neta Scientific Supplier No. 19091GB213
This item is manufactured by Agilent