Learn More
Leica Biosystems HP E27k G5 27-inch 4K UHD USB-C docking monitor
Supplier:  Leica Biosystems 11533663
Encompass_Preferred
The HP E27k G5 27-inch 4K UHD USB-C docking monitor delivers crisp 3840 x 2160 resolution, an IPS panel for wide viewing angles, and integrated docking features-including up to 65W USB-C power delivery, Gigabit Ethernet, and a built-in USB hub-to simplify connections for modern workstations.
  • 27-inch 4K UHD (3840 x 2160) IPS display for detailed visuals.
  • Up to 65W USB-C power delivery to charge compatible laptops.
  • Integrated Gigabit Ethernet and USB hub for docking convenience.
  • Three SuperSpeed USB-A ports, including one charging port.
  • Built-in dual 3W speakers for basic audio playback.
  • 350 nits brightness and 5 ms response time for clear images.
  • Ambient light sensor and eye comfort features to reduce eye strain.
  • 4-way adjustability with ergonomic setup guide for flexible positioning.
Catalog No. NC3366071