Learn More
Neta Scientific HP-INNOWAX 30 M 0.25 MM 0.25UM
SDP
Catalog No. NC1264050
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. NC1264050 Supplier Neta Scientific Supplier No. 19091N133I
This item is manufactured by Agilent