Learn More
IDEX Health & Science Upchurch HP MIXING TEE SS FRIT 10UM
Catalog No. NC1919880
Encompass_Preferred
Idex High-Pressure Tees, Diameter Outer: 0.5 mm, PEEK, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing
Catalog No. NC1919880 Supplier IDEX Health & Science Upchurch Supplier No. U466S
Idex High-Pressure Tees, Diameter Outer: 0.5 mm, PEEK, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing