Learn More
Vici Precision Sampling, Inc. PEEK One-Piece Fingertight Fittings, Blue, 5/pk
Catalog No. NC0854176
Encompass_Preferred
Our PEEK fingertight fittings are available in six colors, for easy identification of tubing lines. They are rated to 350 bar (5000 psi) and can be used in virtually any type of 10-32 thread HPLC fitting detail on the market.
Catalog No. NC0854176 Supplier Vici Precision Sampling, Inc. Supplier No. JR550245
Our PEEK fingertight fittings are available in six colors, for easy identification of tubing lines. They are rated to 350 bar (5000 psi) and can be used in virtually any type of 10-32 thread HPLC fitting detail on the market.