Learn More
Enterprise Technology Solutions HP Thunderbolt 4 280W Ultra G6 Dock
SDP
Catalog No. 5000173765
Encompass_Preferred
Maximize your productivity and enjoy ample connectivity with the Thunderbolt 4 280W Ultra G6 Dock from HP. This dock can support multiple display configurations, including 5K (5120 x 2880) resolution at up to 60 Hz on a single display or up to four simultaneous displays at 4K (3840 x 2160) resolution at up to 60 Hz. 
Catalog No. 50-001-73765 Supplier Enterprise Technology Solutions Supplier No. AW5M5UT
Maximize your productivity and enjoy ample connectivity with the Thunderbolt 4 280W Ultra G6 Dock from HP. This dock can support multiple display configurations, including 5K (5120 x 2880) resolution at up to 60 Hz on a single display or up to four simultaneous displays at 4K (3840 x 2160) resolution at up to 60 Hz. 