Learn More
Sterlitech Corporation HP4750 High Pressure Hose (0.25IDx6ft 0.25MPT)
SDP
Catalog No. 501952343
Encompass_Preferred
HP4750 High Pressure Hose, 6ft length. 
Hose includes one ¼” compression to ¼” male NPT adapter, which is used to connect the hose to the inlet of the HP4750 Cell. The other ¼” tube end of the hose is swaged into the Pressure Bleed Valve.
Catalog No. 50-195-2343 Supplier Sterlitech Corporation Supplier No. 1151427
HP4750 High Pressure Hose, 6ft length. 
Hose includes one ¼” compression to ¼” male NPT adapter, which is used to connect the hose to the inlet of the HP4750 Cell. The other ¼” tube end of the hose is swaged into the Pressure Bleed Valve.