Learn More
New England Biolabs, Inc. HpaII Methyltransferase – 100 units
SDP
Catalog No. 50811632
Encompass
HpaII Methyltransferase recognizes the same sequence as the MspI Methyltransferase, but modifies the internal cytosine residue (C5) of the sequence CCGG.
Catalog No. 50-811-632 Supplier New England Biolabs, Inc. Supplier No. M0214S
HpaII Methyltransferase recognizes the same sequence as the MspI Methyltransferase, but modifies the internal cytosine residue (C5) of the sequence CCGG.