Learn More
Cole-Parmer Thermo Scientific Thermolyne HPA2245MQ Aluminum Hot Plate, 24" x12", 120V, UL/CSA
Catalog No. NC1869564 Shop All Cole-Parmer Products
Encompass
Hot plates feature a sturdy stainless steel case that resists spills and corrosion and supports heavy loads. The 6-1/4'' x 6-1/4'' hot plates have a 20 lb maximum load capacity; all other models have a 40 lb maximum load capacity. Heating surface resists warping and buckling. Embedded heating elements provide better heat transfer and have a longer life than ''open-coil'' designs. Perforated side plates on the 12'' x 12'' and 24'' x 24'' hot plates allow air circulationkeep controls cool to the touch. For added safety, an indicator lights up when power is supplied to the heating element.
Catalog No. NC1869564 Supplier Cole-Parmer Supplier No. 0346000
Hot plates feature a sturdy stainless steel case that resists spills and corrosion and supports heavy loads. The 6-1/4'' x 6-1/4'' hot plates have a 20 lb maximum load capacity; all other models have a 40 lb maximum load capacity. Heating surface resists warping and buckling. Embedded heating elements provide better heat transfer and have a longer life than ''open-coil'' designs. Perforated side plates on the 12'' x 12'' and 24'' x 24'' hot plates allow air circulationkeep controls cool to the touch. For added safety, an indicator lights up when power is supplied to the heating element.