Learn More
Promocell GmbH PromoCell HPAEC-c 500,000 Human Pulmonary Artery Endothelial Cells, cryopreserved
Catalog No. 50305981
Encompass
Primary Human Pulmonary Artery Endothelial Cells (HPAEC) are isolated from human pulmonary arteries (the main pulmonary artery, left and right branches). Since the pulmonary arteries carry blood from the heart to the lungs, they are the only arteries (other than umbilical arteries) that carry deoxygenated blood.
Catalog No. 50-305-981 Supplier Promocell GmbH Supplier No. C12241
Primary Human Pulmonary Artery Endothelial Cells (HPAEC) are isolated from human pulmonary arteries (the main pulmonary artery, left and right branches). Since the pulmonary arteries carry blood from the heart to the lungs, they are the only arteries (other than umbilical arteries) that carry deoxygenated blood.