Learn More
Lonza Walkersville HPAEC – Human Pulmonary Artery Endothelial Cells
Supplier:  Lonza Walkersville CC2530
Encompass
Lonza's Human Pulmonary Artery Endothelial Cells (HPAEC) and media are convenient and easy to use, allowing the researcher to focus on results. Cryopreserved HPAEC cells are shipped in third passage. Proliferating cells are shipped in fourth passage. Cells isolated from Diabetes Type 1 and 2 are also available.Clonetics cells, medium and reagents are quality tested together and guaranteed to give optimum performance as a complete cell system.Benefits:Assured for experimental use up to 15 population doublings All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCharacterized by morphological observation throughout serial passageCertificate of Analysis provided for every cell lot Applications:Hypertension Arteriosclerosis InflammationOncologyDrug screening Content:1 x Cryopreserved ampule of HPAEC containing ≥ 500,000 cells
Catalog No. NC1791225