Learn More
New England Biolabs, Inc. HpaI – 2500 units
SDP
Catalog No. 50811930
Encompass
A restriction endonuclease that recognizes the sequence GTT_^AAC.
Catalog No. 50-811-930 Supplier New England Biolabs, Inc. Supplier No. R0105L
A restriction endonuclease that recognizes the sequence GTT_^AAC.