Learn More
New England Biolabs, Inc. HpaI – 500 units
SDP
Catalog No. 50811931
Encompass
A restriction endonuclease that recognizes the sequence GTT_^AAC.
Catalog No. 50-811-931 Supplier New England Biolabs, Inc. Supplier No. R0105S
A restriction endonuclease that recognizes the sequence GTT_^AAC.