Learn More
New England Biolabs, Inc. HpaII – 2000 units
SDP
Catalog No. 50812081
Encompass
A restriction endonuclease that recognizes the sequence C^CG_G.
Catalog No. 50-812-081 Supplier New England Biolabs, Inc. Supplier No. R0171S
A restriction endonuclease that recognizes the sequence C^CG_G.