Learn More
New England Biolabs, Inc. HpaII – 10000 units
SDP
Catalog No. 50812080
Encompass
A restriction endonuclease that recognizes the sequence C^CG_G.
Catalog No. 50-812-080 Supplier New England Biolabs, Inc. Supplier No. R0171M
A restriction endonuclease that recognizes the sequence C^CG_G.