Learn More
AdipoGen HpARI rec His
SDP
Catalog No. 502562903
Encompass_Preferred
Protein. Heligmosomoides polygyrus protein HpARI aa 17-251 is fused at the C-terminus to a His-tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >90% SDS-PAGE. HpARI is a protein secreted by the mouse parasite Heligmosomoides polygyrus. The mature protein HpARI, containing three predicted Complement Control Protein CCP-like modules also known as Short Consensus Repeats SCRs or sushi-domains, suppresses type 2 allergic immune responses through interference in the interleukin-33 IL-33 pathway. During cell damage, HpARI gains access to the nucleus of necrotic cells, where it binds directly to IL-33 and nuclear DNA, preventing secretion and binding of IL-33 to its receptor. HpARI is a new type of reagent to prevent IL-33-mediated pathology.
Catalog No. 50-256-2903 Supplier AdipoGen Supplier No. AG40B0178C050
Protein. Heligmosomoides polygyrus protein HpARI aa 17-251 is fused at the C-terminus to a His-tag. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >90% SDS-PAGE. HpARI is a protein secreted by the mouse parasite Heligmosomoides polygyrus. The mature protein HpARI, containing three predicted Complement Control Protein CCP-like modules also known as Short Consensus Repeats SCRs or sushi-domains, suppresses type 2 allergic immune responses through interference in the interleukin-33 IL-33 pathway. During cell damage, HpARI gains access to the nucleus of necrotic cells, where it binds directly to IL-33 and nuclear DNA, preventing secretion and binding of IL-33 to its receptor. HpARI is a new type of reagent to prevent IL-33-mediated pathology.