Learn More
Promocell GmbH PromoCell HPASMC-c 500,000 Human Pulmonary Artery Smooth Muscle Cells, cryopreserved
Supplier:  Promocell GmbH C12521
Encompass
Primary Human Pulmonary Artery Smooth Muscle Cells (HPASMC) are isolated from human pulmonary arteries and stain positive for smooth muscle α-actin. They are suited for studying the function of pulmonary arteries under normal and disease conditions (e.g. vasoconstriction induced by alveolar hypoxia). They are also useful for physiological studies of vascular smooth muscle cells.
Catalog No. 50-306-027