Learn More
Lonza Walkersville hPBMC Cryo 10M
Catalog No. 5000496362
Encompass
Lonza s Peripheral Blood Mononuclear Cells (PBMCs) are isolated from peripheral blood by apheresis and density gradient separation Mononuclear cell (MNC) rich cells from leukapheresis are depleted of RBCs and platelets MNC pools differ my donor and may contain on average 50% T cells 20% monocytes 10% B cells 10% NK cells and may contain up to 5% granulocytes They are guaranteed to contain 10 million viable cells per ampoule HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots Cells are collected from healthy donors following IRB protocols A Certificate of Analysis is provided for each cell lot purchased
Catalog No. 50-004-96362 Supplier Lonza Walkersville Supplier No. CC2704
Lonza s Peripheral Blood Mononuclear Cells (PBMCs) are isolated from peripheral blood by apheresis and density gradient separation Mononuclear cell (MNC) rich cells from leukapheresis are depleted of RBCs and platelets MNC pools differ my donor and may contain on average 50% T cells 20% monocytes 10% B cells 10% NK cells and may contain up to 5% granulocytes They are guaranteed to contain 10 million viable cells per ampoule HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots Cells are collected from healthy donors following IRB protocols A Certificate of Analysis is provided for each cell lot purchased