Learn More
Promocell GmbH PromoCell hPC-PL-c 500,000 Human Pericytes, derived from Placenta, cryopreserved
Catalog No. 50306087
Encompass
Pericytes are multipotent mesenchymal-like cells found in association with small blood vessel walls. They are important for angiogenesis, the structural integrity of the microvasculature, and blood flow regulation. However, they can also develop into malignant tumors called hemangiopericytomas.To date, pericytes have demonstrated the ability to differentiate into adipocytes, osteoblasts, fibroblasts, smooth muscle cells, and phagocytes (macrophages). Our hPC-PL are now also available from HLA-typed donors.
Catalog No. 50-306-087 Supplier Promocell GmbH Supplier No. C12980
Pericytes are multipotent mesenchymal-like cells found in association with small blood vessel walls. They are important for angiogenesis, the structural integrity of the microvasculature, and blood flow regulation. However, they can also develop into malignant tumors called hemangiopericytomas.To date, pericytes have demonstrated the ability to differentiate into adipocytes, osteoblasts, fibroblasts, smooth muscle cells, and phagocytes (macrophages). Our hPC-PL are now also available from HLA-typed donors.