Learn More
Aviva Systems Biology Corp Hpcal1 Antibody - C-terminal region (ARP38119_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38119_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hpcal1. It was validated on Western Blot. . Alternate names: NVP-3, Nvp3, VILIP3, Vnsl3, Vsnl3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-718