Learn More
Aviva Systems Biology Corp HPDL Antibody - C-terminal region (ARP60659_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60659_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HPDL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HPDL. Alternate names: 4-HPPD-L, GLOXD1, MGC15668, RP4-534D1.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-118