Learn More
Enzo Life Sciences HPF (1mg). CAS: 359010-69-8
SDP
Supplier:  Enzo Life Sciences ALX620074M001
Encompass_Preferred
Cell permeable fluorescent reagent (Ex(max): 490nm; Em(max): 515nm) for the detection of highly reactive oxygen species (hROS). Immediately reacts with hROS such as hydroxyl radical and peroxynitrite, and the fluorescence intensity greatly increases. In addition, peroxynitrite can be detected in distinction from nitric oxide and superoxide since HPF does not react with nitric oxide, superoxide and hydrogen peroxide. Moreover, HPF is resistant to light-induced autooxidation. HPF does not react with hypochlorite (-OCl) either and thus can be used in combination with APF (ALX-620-075) which detects -OCl to elucidate reliably the roles of -OCl in biological systems such as neutrophils. Moreover, HPF is resistant to light-induced autooxidation. Not for sale in Japan. Alternative name: Hydroxyphenyl fluorescein, 2-[6-(4'-Hydroxy)phenoxy-3H-xanthen-3-on-9-yl]benzoic acid. Formulation: Dissolved in 0.47ml dimethylformamide. Purity: ≥98% (HPLC). Appearance: Pale yellow. Long Term Storage: +4°C.
Catalog No. 50-200-9605