Learn More
Promocell GmbH PromoCell HPF-c 500,000 Human Pulmonary Fibroblasts, cryopreserved
Catalog No. 50306006
Encompass
Primary Human Pulmonary Fibroblasts (HPF) are isolated from human lung tissue. They are often used for studying lung tissue repair, tissue remodeling after injury, or the response after pulmonary inflammation. Fibroblast disorders like pulmonary fibrosis or hypoxia-induced pulmonary hypertension can also be investigated with cultured HPF.
Catalog No. 50-306-006 Supplier Promocell GmbH Supplier No. C12360
Primary Human Pulmonary Fibroblasts (HPF) are isolated from human lung tissue. They are often used for studying lung tissue repair, tissue remodeling after injury, or the response after pulmonary inflammation. Fibroblast disorders like pulmonary fibrosis or hypoxia-induced pulmonary hypertension can also be investigated with cultured HPF.