Learn More
TARGETMOL CHEMICALS INC HPGDS INHIBITOR 1 5MG
SDP
Catalog No. 502581087
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HPGDS inhibitor 1 is a novel and selective inhibitor for Hematopoietic Prostaglandin D Synthase (HPGDS) with an IC50 Value of 0.7 nM. purity: 99%
Catalog No. 50-258-1087 Supplier TARGETMOL CHEMICALS INC Supplier No. T1804
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HPGDS inhibitor 1 is a novel and selective inhibitor for Hematopoietic Prostaglandin D Synthase (HPGDS) with an IC50 Value of 0.7 nM. purity: 99%