Learn More
TARGETMOL CHEMICALS INC HPGDS inhibitor 1 5mg
SDP
Catalog No. NC1624954
Encompass_Preferred
HPGDS inhibitor 1 is a novel and selective inhibitor for Hematopoietic Prostaglandin D Synthase (HPGDS) with an IC50 Value of 0.7 nM.
Catalog No. NC1624954 Supplier TARGETMOL CHEMICALS INC Supplier No. T18045MG
HPGDS inhibitor 1 is a novel and selective inhibitor for Hematopoietic Prostaglandin D Synthase (HPGDS) with an IC50 Value of 0.7 nM.