Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HPGDS inhibitor 2 | 2101626-26-8 | 99.7% | 378.41 g·mol⁻1 | C20H24F2N2O3 | 50MG
SDP

Catalog No. 5000235994
Encompass_Preferred

HPGDS inhibitor 2 is a potent, selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor for research use, with an in vitro IC50 of 9.9 nM. It is supplied as a high-purity solid and includes solubility and formulation data to support biochemical, cellular, and in vivo studies.

  • Potent H-PGDS inhibition (IC50 9.9 nM).
  • High purity (99.7%).
  • Molecular weight 378.41 g·mol⁻1.
  • Chemical formula C20H24F2N2O3.
  • Soluble in DMSO at ≥150 mg/mL for in vitro applications.
  • Formulation-ready for in vivo dosing at ≥2.5 mg/mL in common vehicles.
  • Available as a 50 mg research quantity.

Catalog No. 50-002-35994 Supplier Medchemexpress LLC Supplier No. HY12613450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HPGDS inhibitor 2 is a potent, selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor for research use, with an in vitro IC50 of 9.9 nM. It is supplied as a high-purity solid and includes solubility and formulation data to support biochemical, cellular, and in vivo studies.

  • Potent H-PGDS inhibition (IC50 9.9 nM).
  • High purity (99.7%).
  • Molecular weight 378.41 g·mol⁻1.
  • Chemical formula C20H24F2N2O3.
  • Soluble in DMSO at ≥150 mg/mL for in vitro applications.
  • Formulation-ready for in vivo dosing at ≥2.5 mg/mL in common vehicles.
  • Available as a 50 mg research quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.