Learn More
Medchemexpress LLC HY-126134 5mg Medchemexpress, HPGDS inhibitor 2 CAS:2101626-26-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1261345MG
Encompass_Preferred
Medchemexpress, HY-126134 5mg HPGDS inhibitor 2 CAS:2101626-26-8 HPGDS inhibitor 2 is a highly potent and selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor with an IC 50 of 9.9 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2611