Learn More
New England Biolabs, Inc. HphI – 5000 units
SDP
Catalog No. 50812059
Encompass
A restriction endonuclease that recognizes the sequence GGTGANNNNNNN_N^.
Catalog No. 50-812-059 Supplier New England Biolabs, Inc. Supplier No. R0158L
A restriction endonuclease that recognizes the sequence GGTGANNNNNNN_N^.