Learn More
Selleck Chemical LLC HPI-4 Ciliobrevin A-10mg
SDP
Supplier:  Selleck Chemical LLC S824910MG
Encompass_Preferred
HPI-4 (Ciliobrevin A, Hedgehog Pathway Inhibitor 4) is a hedgehog (Hh) pathway antagonist. It blocks Sonic hedgehog (Shh)-induced Hh pathway activation (IC50 = 7 μM) downstream of Smo.
Catalog No. 50-238-9696