Learn More
Medchemexpress LLC HPK1-IN-3 | 2739844-34-7 | 98.6% | C23H22F4N6O2 | 10MG
SDP
Catalog No. 5000260804
Encompass_Preferred
HPK1-IN-3 is a potent, selective ATP-competitive inhibitor of hematopoietic progenitor kinase 1 (HPK1/MAP4K1) intended for research use. It exhibits sub-nanomolar enzymatic potency and is provided as a high-purity small molecule suitable for biochemical assays and cell-based studies.
  • Potent HPK1 inhibition (IC50 = 0.25 nM).
  • High purity suitable for research applications (98.6%).
  • Suitable for biochemical kinase assays and cell-based pharmacology.
  • Small-molecule format compatible with standard assay preparations.
  • Available in research-scale quantities for assay development.
Catalog No. 50-002-60804 Supplier Medchemexpress LLC Supplier No. HY13856810MG
HPK1-IN-3 is a potent, selective ATP-competitive inhibitor of hematopoietic progenitor kinase 1 (HPK1/MAP4K1) intended for research use. It exhibits sub-nanomolar enzymatic potency and is provided as a high-purity small molecule suitable for biochemical assays and cell-based studies.
  • Potent HPK1 inhibition (IC50 = 0.25 nM).
  • High purity suitable for research applications (98.6%).
  • Suitable for biochemical kinase assays and cell-based pharmacology.
  • Small-molecule format compatible with standard assay preparations.
  • Available in research-scale quantities for assay development.