Learn More
Medchemexpress LLC HPK1-IN-4 | 2739844-28-9 | 434.49 | 100 MG
SDP
Catalog No. 5000350769
Encompass_Preferred
HPK1-IN-4 is an inhibitor of HPK1 (MAPK41), designed as a preclinical immunoresearch tool compound. It exhibits potent activity against HPK1 with a low IC50 value, making it suitable for various research applications targeting the MAPK/ERK pathway. It can be formulated for both in vitro and in vivo studies.
  • Inhibits HPK1 (MAPK41) with high potency
  • Preclinical immunoresearch tool compound
  • Targeted to the MAPK/ERK pathway
  • Suitable for in vitro and in vivo applications
  • Solubility data available for various solvents and formulations
Catalog No. 50-003-50769 Supplier Medchemexpress LLC Supplier No. HY138569100MG
HPK1-IN-4 is an inhibitor of HPK1 (MAPK41), designed as a preclinical immunoresearch tool compound. It exhibits potent activity against HPK1 with a low IC50 value, making it suitable for various research applications targeting the MAPK/ERK pathway. It can be formulated for both in vitro and in vivo studies.
  • Inhibits HPK1 (MAPK41) with high potency
  • Preclinical immunoresearch tool compound
  • Targeted to the MAPK/ERK pathway
  • Suitable for in vitro and in vivo applications
  • Solubility data available for various solvents and formulations