Learn More
Medchemexpress LLC HPK1-IN-4 | 2739844-28-9 | 99.1% | 434.49 | 50 MG
SDP
Catalog No. 5000352357
Encompass_Preferred
HPK1-IN-4 is a potent HPK1 (MAPK41) inhibitor with an IC50 of 0.061 nM. It serves as a preclinical immunotherapy tool compound for research applications.
  • Potent HPK1 (MAPK41) inhibitor
  • IC50 of 0.061 nM
  • Targets MAP4K
  • Affects the MAPK/ERK pathway
  • Preclinical immunotherapy tool compound
Catalog No. 50-003-52357 Supplier Medchemexpress LLC Supplier No. HY13856950MG
HPK1-IN-4 is a potent HPK1 (MAPK41) inhibitor with an IC50 of 0.061 nM. It serves as a preclinical immunotherapy tool compound for research applications.
  • Potent HPK1 (MAPK41) inhibitor
  • IC50 of 0.061 nM
  • Targets MAP4K
  • Affects the MAPK/ERK pathway
  • Preclinical immunotherapy tool compound