Learn More
Medchemexpress LLC HPK1-IN-7 | 2320462-65-3 | 99.3% | 458.47 | 50 MG
SDP
Catalog No. 5000351636
Encompass_Preferred
HPK1-IN-7 is a potent, orally active HPK1 (hematopoietic progenitor kinase 1, MAP4K1) inhibitor (IC50 = 2.6 nM) with excellent family and kinome selectivity. It shows selectivity against IRAK4 (59 nM) and GLK (140 nM). HPK1-IN-7 shows robust efficacy against MC38 syngeneic tumor model in combination with anti-PD1.
  • Potent, orally active HPK1 inhibitor (IC50 = 2.6 nM)
  • Excellent family and kinome selectivity
  • Shows selectivity against IRAK4 (59 nM) and GLK (140 nM)
  • Robust efficacy against MC38 syngeneic tumor model in combination with anti-PD1
Catalog No. 50-003-51636 Supplier Medchemexpress LLC Supplier No. HY13874250MG
HPK1-IN-7 is a potent, orally active HPK1 (hematopoietic progenitor kinase 1, MAP4K1) inhibitor (IC50 = 2.6 nM) with excellent family and kinome selectivity. It shows selectivity against IRAK4 (59 nM) and GLK (140 nM). HPK1-IN-7 shows robust efficacy against MC38 syngeneic tumor model in combination with anti-PD1.
  • Potent, orally active HPK1 inhibitor (IC50 = 2.6 nM)
  • Excellent family and kinome selectivity
  • Shows selectivity against IRAK4 (59 nM) and GLK (140 nM)
  • Robust efficacy against MC38 syngeneic tumor model in combination with anti-PD1