Learn More
Medchemexpress LLC HPK1-IN-7 | 2320462-65-3 | 99.33% | 458.47 | 100 MG
SDP
Catalog No. 5000350577
Encompass_Preferred
HPK1-IN-7 is a potent, orally active hematopoietic progenitor kinase 1 inhibitor with excellent family and kinome selectivity. It demonstrates robust efficacy in tumor models in combination with anti-PD1.
  • Potent and selective inhibitor
  • Orally active
  • Shows selectivity against IRAK4 and GLK
  • Enhances anti-PD1 efficacy in syngeneic tumor models
Catalog No. 50-003-50577 Supplier Medchemexpress LLC Supplier No. HY138742100MG
HPK1-IN-7 is a potent, orally active hematopoietic progenitor kinase 1 inhibitor with excellent family and kinome selectivity. It demonstrates robust efficacy in tumor models in combination with anti-PD1.
  • Potent and selective inhibitor
  • Orally active
  • Shows selectivity against IRAK4 and GLK
  • Enhances anti-PD1 efficacy in syngeneic tumor models