Learn More
Analytical Sales & Services HPLC 48-Position Parallel Synthesis Reaction Block, Gen II
Catalog No. 5000806978
Encompass
Gen II, 48 Well, 4-BOLT, Parallel Synthesis Reaction Block\n• For 12mm OD (2mL) Vials\n• 13mm Well Spacing\n• Includes Vial Rack, Top Cover, PFA Film, Rubber Mats and Screws (vials not included)
Catalog No. 50-008-06978 Supplier Analytical Sales & Services Supplier No. 104480
Gen II, 48 Well, 4-BOLT, Parallel Synthesis Reaction Block\n• For 12mm OD (2mL) Vials\n• 13mm Well Spacing\n• Includes Vial Rack, Top Cover, PFA Film, Rubber Mats and Screws (vials not included)