Learn More
Analytical Sales & Services HPLC 48-Position Photoredox Reaction Block, Gen II
Catalog No. 5000806582
Encompass
4-Bolt, 48-Well Photoredox Reaction Block\n• For 2mL, 12x32mm Vials\n• 13mm Well Spacing\n• Includes Vial Rack, Top Cover, PFA Film, Rubber Mat and Screws (vials not included)\n\n \n\n 
Catalog No. 50-008-06582 Supplier Analytical Sales & Services Supplier No. 101480
4-Bolt, 48-Well Photoredox Reaction Block\n• For 2mL, 12x32mm Vials\n• 13mm Well Spacing\n• Includes Vial Rack, Top Cover, PFA Film, Rubber Mat and Screws (vials not included)\n\n \n\n 