Learn More
WELCH MATERIALS INC Welch MaterialsTM UltisilTM XB-C30, 5μm, 4.6×250mm
Supplier:  WELCH MATERIALS INC H0022331043
Encompass_Preferred
HPLC Column, brand: Ultisil, bonding phase: C30, dimension: 4.6×250mm, particle size: 5 μm, USP: L62, surface area: 320 m2/g, pore size: 120Å, carbon loading: 22%, pH range: 1.5-10.0, endcapped.
Catalog No. 50-196-3713